Assay Details
Target Gene Details
Entrez Gene ID: | 347733 |
Gene Name: | tubulin beta 2B class IIb |
Gene Aliases: |
PMGYSA, bA506K6.1 |
Location: |
Chr.6:3224261-3227734 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBB2B | NM_178012.4 | 4 | 1933 | NP_821080.1 |
| BC063610.1 | 4 | 1836 | AAH63610.1 | |
| BG056085.1 | 1 | 87 | ||
| BX419454.2 | 1 | 532 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv965676 | Chr.6:3224204 - 3226178 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs141781106] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map