Assay Details
Target Gene Details
Entrez Gene ID: | 26248 |
Gene Name: | olfactory receptor family 2 subfamily K member 2 |
Gene Aliases: |
HSHTPCRH06, HTPCRH06, OR2AN1P, OR2AR1P |
Location: |
Chr.9:111326884-111330224 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR2K2 | NM_205859.1 | 1 | 128 | NP_995581.1 |
| XM_011518520.2 | 2 | 296 | XP_011516822.1 | |
| BC111086.1 | 1 | 128 | AAI11087.1 |
More Information
Set Membership: |
|

Genomic Map