Assay Details
Target Gene Details
Entrez Gene ID: | 79228 |
Gene Name: | THO complex 6 |
Gene Aliases: |
BBIS, WDR58, fSAP35 |
Location: |
Chr.16:3024031-3027755 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THOC6 | NM_001142350.1 | NP_001135822.1 | ||
| NM_024339.3 | NP_077315.2 | |||
| AK075330.1 | BAC11552.1 | |||
| AK308960.1 | 2 | 375 | ||
| AK310113.1 | 2 | 387 | ||
| AK314086.1 | ||||
| AL832548.1 | ||||
| BC003118.1 | AAH03118.1 | |||
| BC050674.1 | AAH50674.1 | |||
| BG613317.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3637645 | Chr.16:3021600 - 3057596 on Build GRCh38 | Gain |
|
| nsv509588 | Chr.16:2919880 - 3043776 on Build GRCh38 | Insertion |
|
| nsv833125 | Chr.16:3017124 - 3131817 on Build GRCh38 | Loss |
|
| nsv952906 | Chr.16:2868600 - 3028099 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map