Assay Details
Target Gene Details
Entrez Gene ID: | 57404 |
Gene Name: | cytochrome P450 family 20 subfamily A member 1 |
Gene Aliases: |
CYP-M |
Location: |
Chr.2:203238441-203305840 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP20A1 | NM_177538.2 | 1 | 326 | NP_803882.1 |
| BM141805.1 | 1 | 326 | ||
| BM142090.1 | 1 | 449 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001799 | Chr.2:202908732 - 205615330 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map