Assay Details
Target Gene Details
Entrez Gene ID: | 84268 |
Gene Name: | RPA interacting protein |
Gene Aliases: |
HRIP, RIP |
Location: |
Chr.17:5419185-5433020 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map