Assay Details
Target Gene Details
Entrez Gene ID: | 154796 |
Gene Name: | angiomotin |
Gene Aliases: |
- |
Location: |
Chr.X:112774877-112840946 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMOT | NM_001113490.1 | 10 | 2522 | NP_001106962.1 |
| NM_133265.2 | 11 | 2091 | NP_573572.1 | |
| XM_005262087.1 | 13 | 3267 | XP_005262144.1 | |
| XM_005262090.1 | 10 | 1764 | XP_005262147.1 | |
| XM_011530875.2 | 14 | 3379 | XP_011529177.1 | |
| XM_017029289.1 | 12 | 2295 | XP_016884778.1 | |
| AB028994.3 | 9 | 1295 | BAA83023.3 | |
| AF286598.1 | 11 | 2091 | AAG01851.1 | |
| AY987378.1 | 10 | 2522 | AAY24451.1 | |
| BC130294.1 | 9 | 1512 |
More Information
Set Membership: |
|

Genomic Map