Assay Details
Target Gene Details
Entrez Gene ID: | 221443 |
Gene Name: | O-acyl-ADP-ribose deacylase 1 |
Gene Aliases: |
C6orf130, TARG1, dJ34B21.3 |
Location: |
Chr.6:41066792-41072449 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OARD1 | NM_145063.2 | NP_659500.1 | ||
| AJ420538.1 | ||||
| AK290939.1 | ||||
| AK310688.1 | 4 | 397 | ||
| AK313361.1 | ||||
| BC011709.2 | AAH11709.1 | |||
| BC021579.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv602970 | Chr.6:41070039 - 41258521 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map