Assay Details
Target Gene Details
Entrez Gene ID: | 1490 |
Gene Name: | connective tissue growth factor |
Gene Aliases: |
CCN2, HCS24, IGFBP8, NOV2 |
Location: |
Chr.6:131948176-131951378 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTGF | NM_001901.2 | NP_001892.1 | ||
| AK092280.1 | ||||
| AK125220.1 | ||||
| AK290884.1 | ||||
| AK315207.1 | ||||
| AY395801.1 | AAQ95223.1 | |||
| AY550024.1 | AAS55639.1 | |||
| BC087839.1 | AAH87839.1 | |||
| BT019794.1 | AAV38597.1 | |||
| BT019795.1 | AAV38598.1 | |||
| CR541734.1 | CAG46534.1 | |||
| CR541759.1 | CAG46559.1 | |||
| DA552292.1 | ||||
| M92934.1 | AAA91279.1 | |||
| X78947.1 | CAA55544.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7968 | Chr.6:131596032 - 131999959 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map