Assay Details
Target Gene Details
Entrez Gene ID: | 5045 |
Gene Name: | furin, paired basic amino acid cleaving enzyme |
Gene Aliases: |
FUR, PACE, PCSK3, SPC1 |
Location: |
Chr.15:90868592-90883458 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FURIN | NM_001289823.1 | NP_001276752.1 | ||
| NM_001289824.1 | NP_001276753.1 | |||
| NM_002569.3 | NP_002560.1 | |||
| AL110107.1 | ||||
| BC012181.1 | AAH12181.1 | |||
| S65442.1 | AAB28140.1 | |||
| X17094.1 | CAA34948.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952028 | Chr.15:90875371 - 90890970 on Build GRCh38 | Deletion |
|
| nsv515661 | Chr.15:90866779 - 90981967 on Build GRCh38 | Loss |
|
| nsv833089 | Chr.15:90859910 - 90962961 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map