Assay Details
Target Gene Details
Entrez Gene ID: | 79092 |
Gene Name: | caspase recruitment domain family member 14 |
Gene Aliases: |
BIMP2, CARMA2, PRP, PSORS2, PSS1 |
Location: |
Chr.17:80169992-80209331 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 23 - Intron 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARD14 | NM_024110.4 | NP_077015.2 | ||
| NR_047566.1 | ||||
| XM_011525212.1 | XP_011523514.1 | |||
| XM_011525213.1 | XP_011523515.1 | |||
| XM_011525215.1 | XP_011523517.1 | |||
| XM_011525216.1 | XP_011523518.1 | |||
| XM_011525217.1 | XP_011523519.1 | |||
| XM_011525218.2 | XP_011523520.1 | |||
| XM_017025057.1 | XP_016880546.1 | |||
| AF322642.1 | AAG53403.1 | |||
| AK002138.1 | ||||
| AK091123.1 | ||||
| AX746790.1 | ||||
| AY032927.1 | AAK54453.1 |
Target Gene Details
Entrez Gene ID: | 6448 |
Gene Name: | N-sulfoglucosamine sulfohydrolase |
Gene Aliases: |
HSS, MPS3A, SFMD |
Location: |
Chr.17:80200668-80220400 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SGSH |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066606 | Chr.17:79955666 - 80224006 on Build GRCh38 | Gain |
|
| nsv469591 | Chr.17:80085322 - 80293486 on Build GRCh38 | Loss |
|
| nsv457968 | Chr.17:80205094 - 80292960 on Build GRCh38 | Loss |
|
| nsv576130 | Chr.17:80184196 - 80220032 on Build GRCh38 | Loss |
|
| nsv833562 | Chr.17:80114523 - 80301339 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map