Assay Details
Target Gene Details
Entrez Gene ID: | 2562 |
Gene Name: | gamma-aminobutyric acid type A receptor beta3 subunit |
Gene Aliases: |
ECA5 |
Location: |
Chr.15:26543546-26773788 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GABRB3 | NM_000814.5 | 9 | 3210 | NP_000805.1 |
| NM_001191320.1 | 7 | 3008 | NP_001178249.1 | |
| NM_001191321.2 | 7 | 3058 | NP_001178250.1 | |
| NM_001278631.1 | 10 | 3306 | NP_001265560.1 | |
| NM_021912.4 | 9 | 3160 | NP_068712.1 | |
| XM_011521428.2 | 8 | 3193 | XP_011519730.1 | |
| AK056018.1 | 1 | 581 | ||
| AK095011.1 | 1 | 1361 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv144n21 | Chr.15:26513916 - 26649934 on Build GRCh38 | Gain |
|
| nsv568600 | Chr.15:26493638 - 26552355 on Build GRCh38 | Gain |
|
| nsv568598 | Chr.15:26082319 - 26575978 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map