Assay Details
Target Gene Details
Entrez Gene ID: | 285172 |
Gene Name: | family with sequence similarity 126 member B |
Gene Aliases: |
HYCC2 |
Location: |
Chr.2:200973718-201071671 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM126B | NM_001321618.1 | 11 | 2500 | NP_001308547.1 |
| NM_001321619.1 | 12 | 2711 | NP_001308548.1 | |
| NM_001321621.1 | 11 | 2613 | NP_001308550.1 | |
| NM_001321622.1 | 13 | 2829 | NP_001308551.1 | |
| NM_001321623.1 | 13 | 2766 | NP_001308552.1 | |
| NM_001321624.1 | 12 | 2668 | NP_001308553.1 | |
| NM_001321625.1 | 13 | 2879 | NP_001308554.1 | |
| NM_001321626.1 | 13 | 2899 | NP_001308555.1 | |
| NM_001321627.1 | 14 | 2822 | NP_001308556.1 | |
| NM_001321628.1 | 14 | 2935 | NP_001308557.1 | |
| NM_001321629.1 | 13 | 2654 | NP_001308558.1 | |
| NM_173822.4 | 12 | 2598 | NP_776183.1 | |
| XM_006712443.3 | 12 | 2781 | XP_006712506.1 | |
| XM_011511002.2 | 13 | 2796 | XP_011509304.1 | |
| XM_017003879.1 | 14 | 2977 | XP_016859368.1 | |
| XM_017003880.1 | 13 | 3391 | XP_016859369.1 | |
| XM_017003881.1 | 12 | 2659 | XP_016859370.1 | |
| XM_017003882.1 | 14 | 2878 | XP_016859371.1 | |
| XM_017003883.1 | 13 | 3053 | XP_016859372.1 | |
| AK001843.1 | ||||
| AK027168.1 | 1 | 473 | ||
| AK097111.1 | 5 | 1841 | ||
| AL832234.1 | 11 | 2457 | ||
| AL832346.1 | 14 | 2817 | ||
| AL833403.1 | 12 | 2582 | ||
| BC039295.1 | 12 | 2596 | AAH39295.1 |
More Information
Set Membership: |
|

Genomic Map