Assay Details
Target Gene Details
Entrez Gene ID: | 84337 |
Gene Name: | elongation factor 1 homolog |
Gene Aliases: |
ELF1 |
Location: |
Chr.19:11553043-11559236 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELOF1 | NM_032377.3 | NP_115753.1 | ||
| AK097850.1 | ||||
| BG718042.1 | 1 | 110 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509721 | Chr.19:11525216 - 11691390 on Build GRCh38 | Insertion |
|
| nsv519802 | Chr.19:11236817 - 11578343 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map