Assay Details
Target Gene Details
Entrez Gene ID: | 290 |
Gene Name: | alanyl aminopeptidase, membrane |
Gene Aliases: |
APN, CD13, GP150, LAP1, P150, PEPN |
Location: |
Chr.15:89784895-89814852 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANPEP | NM_001150.2 | 8 | 1680 | NP_001141.2 |
| XM_005254892.4 | 8 | 1688 | XP_005254949.1 | |
| XM_011521473.1 | 8 | 1831 | XP_011519775.1 | |
| AB209918.1 | 8 | 1676 | BAD93155.1 | |
| AK298134.1 | 2 | 150 | ||
| AK298242.1 | ||||
| AK298339.1 | 3 | 261 | ||
| AK300946.1 | 4 | 411 | ||
| BC058928.1 | 7 | 1481 | AAH58928.1 | |
| M22324.1 | 7 | 1508 | AAA51719.1 | |
| X13276.1 | 7 | 1412 | CAA31640.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519689 | Chr.15:89803630 - 89806327 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map