Assay Details
Target Gene Details
Entrez Gene ID: | 8760 |
Gene Name: | CDP-diacylglycerol synthase 2 |
Gene Aliases: |
- |
Location: |
Chr.20:5126761-5197887 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDS2 | NM_003818.3 | NP_003809.1 | ||
| XM_006723660.1 | XP_006723723.1 | |||
| XM_011529390.1 | XP_011527692.1 | |||
| AK001414.1 | ||||
| AK027695.1 | ||||
| AK091045.1 | ||||
| AK304763.1 | ||||
| AK315489.1 | ||||
| BC025751.1 | AAH25751.1 | |||
| BP353230.1 | ||||
| Y16521.1 | CAA76270.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428375 | Chr.20:4987311 - 5144079 on Build GRCh38 | Loss |
|
| nsv1066860 | Chr.20:4973849 - 5652866 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map