Assay Details
Target Gene Details
Entrez Gene ID: | 6666 |
Gene Name: | SRY-box 12 |
Gene Aliases: |
SOX22 |
Location: |
Chr.20:325571-330228 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOX12 | NM_006943.3 | 1 | 106 | NP_008874.2 |
| BC067361.1 | 1 | 118 | AAH67361.1 | |
| HY104751.1 | 1 | 99 | ||
| U35612.1 | 1 | 82 | AAB69627.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1133498 | Chr.20:325656 - 328856 on Build GRCh38 | Deletion |
|
| dgv4219n100 | Chr.20:80652 - 461103 on Build GRCh38 | Gain |
|
| esv3893263 | Chr.20:216521 - 434561 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs563163591] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map