Assay Details
Target Gene Details
Entrez Gene ID: | 55272 |
Gene Name: | IMP3, U3 small nucleolar ribonucleoprotein |
Gene Aliases: |
BRMS2, C15orf12, MRPS4 |
Location: |
Chr.15:75639085-75640323 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IMP3 | NM_018285.3 | 1 | 605 | NP_060755.1 |
| AA365493.1 | ||||
| AK001830.1 | 1 | 497 | BAA91929.1 | |
| AK057531.1 | 2 | 1394 | ||
| AK223121.1 | 1 | 506 | BAD96841.1 | |
| AL136913.1 | 1 | 494 | CAB66847.1 | |
| AY050497.1 | 1 | 492 | AAL06639.1 | |
| BC006487.1 | 1 | 493 | AAH06487.1 | |
| BC011745.2 | 1 | 476 | AAH11745.1 | |
| BM970419.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2749895 | Chr.15:75415580 - 76472296 on Build GRCh38 | Deletion |
|
| esv2749894 | Chr.15:75326873 - 75749895 on Build GRCh38 | Deletion |
|
| nsv527186 | Chr.15:75632102 - 75815389 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map