Assay Details
Target Gene Details
Entrez Gene ID: | 221060 |
Gene Name: | chromosome 10 open reading frame 111 |
Gene Aliases: |
bA455B2.4 |
Location: |
Chr.10:15095385-15097319 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C10orf111 | NM_153244.1 | 2 | 698 | NP_694976.1 |
| AK314131.1 | 1 | 903 | ||
| BC029034.1 | 2 | 698 | AAH29034.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831794 | Chr.10:14993658 - 15148891 on Build GRCh38 | Loss |
|

Genomic Map