Assay Details
Target Gene Details
Entrez Gene ID: | 27012 |
Gene Name: | potassium voltage-gated channel modifier subfamily V member 1 |
Gene Aliases: |
HNKA, KCNB3, KV2.3, KV8.1 |
Location: |
Chr.8:109963645-109975847 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNV1 | NM_014379.3 | 4 | 3170 | NP_055194.1 |
| AF167082.2 | 3 | 2478 | AAD48031.2 | |
| AK308123.1 | 4 | 3163 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831421 | Chr.8:109862405 - 110026066 on Build GRCh38 | Gain |
|
| nsv6347 | Chr.8:109965914 - 110010961 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map