Assay Details
Target Gene Details
Entrez Gene ID: | 7453 |
Gene Name: | tryptophanyl-tRNA synthetase |
Gene Aliases: |
GAMMA-2, IFI53, IFP53 |
Location: |
Chr.14:100333788-100376343 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WARS | NM_004184.3 | NP_004175.2 | ||
| NM_173701.1 | NP_776049.1 | |||
| NM_213645.1 | NP_998810.1 | |||
| NM_213646.1 | NP_998811.1 | |||
| XM_005268044.3 | XP_005268101.1 | |||
| XM_006720249.3 | XP_006720312.1 | |||
| XM_011537133.2 | XP_011535435.1 | |||
| XM_011537135.2 | XP_011535437.1 | |||
| XM_011537136.2 | XP_011535438.1 | |||
| XM_017021627.1 | XP_016877116.1 | |||
| XM_017021628.1 | XP_016877117.1 | |||
| XM_017021629.1 | XP_016877118.1 | |||
| AK025487.1 | ||||
| AK056100.1 | ||||
| AK130698.1 | ||||
| AK291141.1 | ||||
| BC017489.1 | AAH17489.1 | |||
| BC095453.1 | AAH95453.1 | |||
| BX248006.1 | CAD62335.1 | |||
| M61715.1 | AAA61298.1 | |||
| M77804.1 | AAA67324.1 | |||
| X59892.1 | CAA42545.1 | |||
| X62570.1 | CAA44450.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832875 | Chr.14:100192592 - 100368905 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map