Assay Details
Target Gene Details
Entrez Gene ID: | 100130331 |
Gene Name: | POTE ankyrin domain family, member F pseudogene |
Gene Aliases: |
- |
Location: |
Chr.1:237862175-237928321 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100130331 | NR_027247.2 | |||
| AL122093.1 |
Target Gene Details
Entrez Gene ID: | 57829 |
Gene Name: | zona pellucida glycoprotein 4 |
Gene Aliases: |
ZBP, ZP1, ZPB, Zp-4 |
Location: |
Chr.1:237877864-237890922 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZP4 | NM_021186.4 | NP_067009.1 | ||
| AK314151.1 | ||||
| BC069521.1 | AAH69521.1 | |||
| BE439656.1 | ||||
| DV080723.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34468 | Chr.1:237849470 - 238562659 on Build GRCh38 | Gain |
|
| esv3589241 | Chr.1:237840642 - 238052824 on Build GRCh38 | Gain |
|
| esv2759008 | Chr.1:237778494 - 238604802 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map