Assay Details
Target Gene Details
Entrez Gene ID: | 23603 |
Gene Name: | coronin 1C |
Gene Aliases: |
HCRNN4 |
Location: |
Chr.12:108645109-108731550 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CORO1C | NM_001105237.2 | 5 | 755 | NP_001098707.1 |
| NM_001276471.1 | 5 | 773 | NP_001263400.1 | |
| NM_014325.3 | 5 | 674 | NP_055140.1 | |
| XM_017019121.1 | 6 | 1503 | XP_016874610.1 | |
| XM_017019122.1 | 5 | 1080 | XP_016874611.1 | |
| XM_017019123.1 | 6 | 1133 | XP_016874612.1 | |
| AB030656.1 | 5 | 643 | BAA83077.1 | |
| AB209909.1 | 5 | 629 | BAD93146.1 | |
| AK023205.1 | 5 | 644 | ||
| AK096363.1 | 5 | 773 | ||
| AK223073.1 | 5 | 642 | BAD96793.1 | |
| AK297463.1 | 5 | 507 | ||
| AK304839.1 | 5 | 410 | ||
| AK311315.1 | ||||
| AK316065.1 | 5 | 470 | ||
| AK316504.1 | 4 | 395 | ||
| AL162070.1 | 5 | 612 | CAB82406.1 | |
| AM849477.1 | 5 | 648 | ||
| AM849478.1 | 5 | 755 | ||
| BC002342.2 | 5 | 628 | AAH02342.1 |
More Information
Set Membership: |
|

Genomic Map