Assay Details
Target Gene Details
Entrez Gene ID: | 9616 |
Gene Name: | ring finger protein 7 |
Gene Aliases: |
CKBBP1, ROC2, SAG |
Location: |
Chr.3:141738209-141746803 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF7 | NM_001201370.1 | 2 | 959 | NP_001188299.1 |
| NM_014245.4 | 3 | 1007 | NP_055060.1 | |
| NM_183237.2 | 3 | 1000 | NP_899060.1 | |
| NR_037702.1 | 4 | 1271 | ||
| NR_037703.1 | 4 | 1264 | ||
| BF439211.1 | 2 | 225 | ||
| CA415974.1 |
More Information
Set Membership: |
|

Genomic Map