Assay Details
Target Gene Details
Entrez Gene ID: | 151636 |
Gene Name: | deltex E3 ubiquitin ligase 3L |
Gene Aliases: |
BBAP |
Location: |
Chr.3:122564338-122575203 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DTX3L | NM_138287.3 | 3 | 1197 | NP_612144.1 |
| AF484416.1 | 3 | 1108 | AAL90859.1 | |
| AK125086.1 | 3 | 1197 | ||
| AL833598.1 | 3 | 1130 | ||
| AY225123.1 | 3 | 1242 | AAP57517.1 | |
| AY780792.1 | AAV98362.1 | |||
| BC042191.1 | 3 | 1116 | AAH42191.1 | |
| BC060509.1 | 3 | 1139 | AAH60509.1 | |
| BX648267.1 | 1 | 621 | ||
| BX648645.1 | 3 | 950 |
More Information
Set Membership: |
|

Genomic Map