Assay Details
Target Gene Details
Entrez Gene ID: | 56882 |
Gene Name: | CDC42 small effector 1 |
Gene Aliases: |
SCIP1, SPEC1 |
Location: |
Chr.1:151050971-151059649 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC42SE1 | NM_001038707.1 | 6 | 1019 | NP_001033796.1 |
| NM_020239.3 | 5 | 883 | NP_064624.1 | |
| XM_017001847.1 | 6 | 963 | XP_016857336.1 | |
| AF187845.2 | 5 | 808 | AAF87597.1 | |
| AF286041.1 | 4 | 443 | AAF97248.1 | |
| AF286592.1 | 4 | 1360 | AAG17723.1 | |
| BC012796.2 | 4 | 627 | AAH12796.2 | |
| BC041604.1 | 5 | 845 | AAH41604.1 | |
| BX339337.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map