Assay Details
Target Gene Details
Entrez Gene ID: | 1346 |
Gene Name: | cytochrome c oxidase subunit 7A1 |
Gene Aliases: |
COX7A, COX7AH, COX7AM |
Location: |
Chr.19:36150922-36152869 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX7A1 | NM_001864.3 | NP_001855.1 | ||
| BC002757.2 | AAH02757.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv579425 | Chr.19:36152793 - 36194851 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs142294636] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map