Assay Details
Target Gene Details
Entrez Gene ID: | 9209 |
Gene Name: | LRR binding FLII interacting protein 2 |
Gene Aliases: |
HUFI-2 |
Location: |
Chr.3:37052626-37176817 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 30 - Exon 30 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834663 | Chr.3:36966258 - 37148914 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map