Assay Details
Target Gene Details
Entrez Gene ID: | 7225 |
Gene Name: | transient receptor potential cation channel subfamily C member 6 |
Gene Aliases: |
FSGS2, TRP6 |
Location: |
Chr.11:101451470-101584018 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPC6 | NM_004621.5 | NP_004612.2 | ||
| XM_011542968.2 | XP_011541270.1 | |||
| XM_017018221.1 | XP_016873710.1 | |||
| XM_017018222.1 | XP_016873711.1 | |||
| AF080394.2 | AAC63289.2 | |||
| AJ006276.1 | CAA06943.1 | |||
| AJ271066.1 | CAC01684.1 | |||
| AJ271067.1 | CAC01685.1 | |||
| AJ271068.1 | CAC01686.1 | |||
| AK001960.1 | ||||
| AK027769.1 | ||||
| BC093658.1 | AAH93658.1 | |||
| BC093660.1 | AAH93660.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv468851 | Chr.11:101397716 - 101460076 on Build GRCh38 | Gain |
|
| nsv556131 | Chr.11:101115046 - 101609286 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72984209] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map