Assay Details
Target Gene Details
Entrez Gene ID: | 64755 |
Gene Name: | chromosome 16 open reading frame 58 |
Gene Aliases: |
RUS |
Location: |
Chr.16:31489464-31508419 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C16orf58 | NM_022744.3 | 13 | 2027 | NP_073581.2 |
| XM_017023570.1 | 11 | 1858 | XP_016879059.1 | |
| AK000679.1 | 9 | 1448 | ||
| AK023930.1 | 13 | 1993 | BAB14730.1 | |
| AK092580.1 | 4 | 1655 | BAC03918.1 | |
| AK222913.1 | 13 | 1993 | BAD96633.1 | |
| AK304608.1 | ||||
| BC000822.1 | AAH00822.1 | |||
| BC009308.2 | AAH09308.1 |
Target Gene Details
Entrez Gene ID: | 6524 |
Gene Name: | solute carrier family 5 member 2 |
Gene Aliases: |
SGLT2 |
Location: |
Chr.16:31482535-31490770 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC5A2 | NM_003041.3 | NP_003032.1 | ||
| NR_130783.1 | ||||
| XM_006721072.3 | 13 | 2425 | XP_006721135.2 | |
| AI222550.1 | ||||
| AK054830.1 | 3 | 1959 | ||
| BC131542.1 | ||||
| M95549.1 | AAA36608.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1060240 | Chr.16:31484552 - 31570129 on Build GRCh38 | Gain |
|
| nsv833194 | Chr.16:31346858 - 31517224 on Build GRCh38 | Loss |
|
| nsv482193 | Chr.16:31483118 - 31490770 on Build GRCh38 | Gain |
|
| esv3570465 | Chr.16:31450067 - 31915765 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map