Assay Details
Target Gene Details
Entrez Gene ID: | 7180 |
Gene Name: | cysteine rich secretory protein 2 |
Gene Aliases: |
CRISP-2, CT36, GAPDL5, TPX1, TSP1 |
Location: |
Chr.6:49676665-49714198 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRISP2 | NM_001142407.2 | 9 | 1256 | NP_001135879.1 |
| NM_001142408.2 | 10 | 1313 | NP_001135880.1 | |
| NM_001142417.2 | 10 | 1293 | NP_001135889.1 | |
| NM_001142435.2 | 9 | 1276 | NP_001135907.1 | |
| NM_001261822.1 | 9 | 1282 | NP_001248751.1 | |
| NM_003296.3 | 10 | 1319 | NP_003287.1 | |
| XM_005249349.1 | 11 | 1398 | XP_005249406.1 | |
| XM_005249350.1 | 10 | 1356 | XP_005249407.1 | |
| XM_005249351.3 | 10 | 1376 | XP_005249408.1 | |
| XM_005249352.3 | 10 | 1367 | XP_005249409.1 | |
| XM_005249353.3 | 11 | 1403 | XP_005249410.1 | |
| XM_005249356.1 | 11 | 1401 | XP_005249413.1 | |
| XM_011514841.1 | 11 | 1733 | XP_011513143.1 | |
| XM_011514842.1 | 10 | 1696 | XP_011513144.1 | |
| XM_011514843.2 | XP_011513145.1 | |||
| AK128009.1 | 1 | 2249 | ||
| AK223337.1 | 10 | 1256 | BAD97057.1 | |
| AY289796.1 | 10 | 1263 | AAP41200.1 | |
| AY292862.1 | 10 | 1358 | AAP44113.1 | |
| AY292863.1 | 9 | 1257 | AAP44114.1 | |
| AY773479.1 | 10 | 1299 | AAV48552.1 | |
| BC022011.2 | 10 | 1266 | AAH22011.1 | |
| BC107707.1 | 9 | 1213 | AAI07708.1 | |
| BI831679.1 | 4 | 668 | ||
| DB524518.1 | 1 | 385 | ||
| M25532.1 | 10 | 1242 | AAA61220.1 | |
| X95239.1 | 10 | 1293 | CAA64526.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3608949 | Chr.6:49690413 - 49880115 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map