Assay Details
Target Gene Details
Entrez Gene ID: | 5913 |
Gene Name: | receptor associated protein of the synapse |
Gene Aliases: |
CMS11, CMS4C, FADS, RAPSYN, RNF205 |
Location: |
Chr.11:47437757-47449178 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAPSN | NM_005055.4 | 7 | 1323 | NP_005046.2 |
| NM_032645.4 | 5 | 1146 | NP_116034.2 | |
| XM_005253042.3 | 6 | 1227 | XP_005253099.1 | |
| XM_005253043.3 | 6 | 1158 | XP_005253100.1 | |
| XM_011520252.1 | 7 | 1366 | XP_011518554.1 | |
| AF449218.1 | 7 | 1323 | AAL86639.1 | |
| AK310002.1 | 5 | 1104 | ||
| BC004196.2 | 5 | 1111 | AAH04196.1 | |
| KU178300.1 | 5 | 932 | ||
| KU178301.1 | 7 | 1245 | ||
| KU178302.1 | 7 | 1109 | ||
| KU178303.1 | 6 | 1055 | ||
| KU178304.1 | 6 | 986 | ||
| Z33905.1 | 7 | 1316 | CAA83954.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832141 | Chr.11:47311629 - 47499441 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map