Assay Details
Target Gene Details
Entrez Gene ID: | 81565 |
Gene Name: | nudE neurodevelopment protein 1 like 1 |
Gene Aliases: |
EOPA, MITAP1, NDE1L1, NDE2, NUDEL |
Location: |
Chr.17:8435852-8472744 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDEL1 | NM_001025579.2 | 6 | 789 | NP_001020750.1 |
| NM_030808.4 | 6 | 789 | NP_110435.1 | |
| XM_011524013.1 | 6 | 780 | XP_011522315.1 | |
| XM_017025183.1 | 7 | 969 | XP_016880672.1 | |
| XM_017025184.1 | 7 | 969 | XP_016880673.1 | |
| XM_017025185.1 | 6 | 838 | XP_016880674.1 | |
| XM_017025186.1 | 7 | 969 | XP_016880675.1 | |
| XM_017025187.1 | 6 | 838 | XP_016880676.1 | |
| XM_017025188.1 | 7 | 969 | XP_016880677.1 | |
| AF182078.1 | 6 | 716 | AAG43425.1 | |
| AF217798.3 | 6 | 780 | AAF24516.3 | |
| AK056014.1 | 6 | 787 | ||
| AK299559.1 | 7 | 763 | ||
| AL832648.1 | 6 | 723 | CAD89957.3 | |
| AY004871.1 | 6 | 715 | AAF97497.1 | |
| BC026101.2 | 6 | 727 | AAH26101.1 | |
| CX166766.1 | 6 | 718 | ||
| DA256375.1 | 2 | 130 |
More Information
Set Membership: |
|

Genomic Map