Assay Details
Target Gene Details
Entrez Gene ID: | 338339 |
Gene Name: | C-type lectin domain family 4 member D |
Gene Aliases: |
CD368, CLEC-6, CLEC6, CLECSF8, MCL, MPCL |
Location: |
Chr.12:8513492-8522366 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC4D | NM_080387.4 | 4 | 506 | NP_525126.2 |
| XM_011520632.2 | 5 | 475 | XP_011518934.1 | |
| XM_011520633.2 | 4 | 501 | XP_011518935.1 | |
| AF411850.1 | 4 | 376 | AAL37713.1 | |
| AK314071.1 | 4 | 493 | ||
| AY115592.1 | 4 | 497 | AAM75389.1 | |
| AY297446.1 | 4 | 505 | AAQ63173.1 | |
| AY486482.1 | 4 | 330 | AAS59161.1 | |
| AY486483.1 | 4 | 330 | AAS59162.1 | |
| BC032313.1 | 4 | 506 | AAH32313.1 | |
| BI909298.1 | 4 | 455 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain |
|
| nsv1035270 | Chr.12:8466966 - 8521466 on Build GRCh38 | Gain |
|
| nsv557342 | Chr.12:8446801 - 8621931 on Build GRCh38 | Loss |
|
| dgv1362n100 | Chr.12:8296504 - 8639973 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map