Assay Details
Target Gene Details
Entrez Gene ID: | 5454 |
Gene Name: | POU class 3 homeobox 2 |
Gene Aliases: |
BRN2, N-Oct3, OCT7, OTF-7, OTF7, POUF3, brn-2, oct-7 |
Location: |
Chr.6:98834704-98838790 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POU3F2 | NM_005604.3 | 1 | 2512 | NP_005595.2 |
| BC051699.1 | 1 | 2481 | AAH51699.2 | |
| BQ423615.1 | 1 | 394 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv604253 | Chr.6:98409521 - 99182162 on Build GRCh38 | Gain |
|
| nsv823787 | Chr.6:98822673 - 98854658 on Build GRCh38 | Gain |
|
| nsv1029536 | Chr.6:98422726 - 99818247 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map