Assay Details
Target Gene Details
Entrez Gene ID: | 6619 |
Gene Name: | small nuclear RNA activating complex polypeptide 3 |
Gene Aliases: |
PTFbeta, SNAP50 |
Location: |
Chr.9:15422696-15466751 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNAPC3 | NM_001039697.1 | NP_001034786.1 | ||
| XM_017015056.1 | XP_016870545.1 | |||
| XM_017015057.1 | XP_016870546.1 | |||
| AK293310.1 | ||||
| BC014985.1 | AAH14985.1 | |||
| BC108743.1 | AAI08744.1 | |||
| BC121011.1 | ||||
| BC121012.1 | ||||
| U66413.1 | AAD09214.1 | |||
| U71300.1 | AAC50948.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517975 | Chr.9:15286448 - 15534072 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map