Assay Details
Target Gene Details
Entrez Gene ID: | 10826 |
Gene Name: | fatty acid hydroxylase domain containing 2 |
Gene Aliases: |
C5orf4 |
Location: |
Chr.5:154817018-154850685 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAXDC2 | NM_032385.4 | 9 | 1539 | NP_115761.2 |
| XM_005268358.2 | 6 | 1184 | XP_005268415.1 | |
| XM_006714753.2 | 10 | 1914 | XP_006714816.1 | |
| XM_017008965.1 | 10 | 1939 | XP_016864454.1 | |
| XM_017008966.1 | 7 | 1214 | XP_016864455.1 | |
| AF159165.1 | 1 | 1545 | AAF22611.1 | |
| AK023820.1 | 8 | 2330 | ||
| AK074245.1 | 3 | 684 | ||
| AK127944.1 | 3 | 2873 | ||
| AL359653.1 | 1 | 87 | ||
| AL359655.1 | ||||
| BC004506.2 | 7 | 1236 | AAH04506.2 | |
| BC007216.1 | 9 | 1507 | AAH07216.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830521 | Chr.5:154768007 - 154864706 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map