Assay Details
Target Gene Details
Entrez Gene ID: | 7319 |
Gene Name: | ubiquitin conjugating enzyme E2 A |
Gene Aliases: |
HHR6A, MRXS30, MRXSN, RAD6A, UBC2 |
Location: |
Chr.X:119574467-119584429 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2A | NM_001282161.1 | 1 | 124 | NP_001269090.1 |
| NM_003336.3 | 1 | 124 | NP_003327.2 | |
| NM_181762.2 | 1 | 124 | NP_861427.1 | |
| AK223045.1 | BAD96765.1 | |||
| AK223064.1 | BAD96784.1 | |||
| AK297696.1 | ||||
| AK301675.1 | ||||
| AK313092.1 | ||||
| AL701289.1 | ||||
| BC010175.2 | AAH10175.1 | |||
| D28459.1 | BAA05825.1 | |||
| DA068506.1 | ||||
| DA599154.1 | 1 | 124 | ||
| M74524.1 | AAA35981.1 |
More Information
Set Membership: |
|

Genomic Map