Assay Details
Target Gene Details
Entrez Gene ID: | 317716 |
Gene Name: | BPI fold containing family A member 4, pseudogene |
Gene Aliases: |
BASE |
Location: |
Chr.20:33193605-33210462 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BPIFA4P | NR_026760.1 | |||
| AY180924.1 | AAO17728.1 | |||
| BC128240.1 | ||||
| BC128241.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056411 | Chr.20:32469234 - 33355029 on Build GRCh38 | Loss |
|
| esv2751912 | Chr.20:33179619 - 33248657 on Build GRCh38 | Loss |
|
| nsv1060241 | Chr.20:33172902 - 33244962 on Build GRCh38 | Loss |
|
| nsv458964 | Chr.20:33152998 - 33229798 on Build GRCh38 | Loss |
|

Genomic Map