Assay Details
Target Gene Details
Entrez Gene ID: | 11168 |
Gene Name: | PC4 and SFRS1 interacting protein 1 |
Gene Aliases: |
DFS70, LEDGF, PAIP, PSIP2, p52, p75 |
Location: |
Chr.9:15464066-15511019 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSIP1 | NM_001128217.2 | 14 | 1632 | NP_001121689.1 |
| NM_033222.4 | 14 | 1635 | NP_150091.2 | |
| AF063020.1 | 14 | 1601 | AAC25167.1 | |
| AF098483.1 | 13 | 1315 | AAC97946.1 | |
| AF432220.1 | 14 | 1583 | AAL99926.1 | |
| BX649155.1 | 4 | 3017 | ||
| U94319.1 | 8 | 754 | AAB52589.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517975 | Chr.9:15286448 - 15534072 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map