Assay Details
Target Gene Details
Entrez Gene ID: | 283284 |
Gene Name: | immunoglobulin superfamily member 22 |
Gene Aliases: |
IGFN2 |
Location: |
Chr.11:18704305-18726939 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGSF22 | NM_173588.3 | 11 | 1494 | NP_775859.3 |
| XM_011520030.2 | 9 | 4398 | XP_011518332.1 | |
| XM_011520031.2 | 9 | 4398 | XP_011518333.1 | |
| XM_017017607.1 | 10 | 4289 | XP_016873096.1 | |
| XM_017017608.1 | 10 | 2312 | XP_016873097.1 | |
| AK095113.1 | 11 | 1494 | BAC04488.1 | |
| BC115397.1 | 11 | 1445 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047371 | Chr.11:18709926 - 18741490 on Build GRCh38 | Gain |
|
| nsv553633 | Chr.11:18290154 - 19036904 on Build GRCh38 | Gain |
|
| nsv525192 | Chr.11:18701244 - 18771813 on Build GRCh38 | Loss |
|
| nsv525452 | Chr.11:18714400 - 18780956 on Build GRCh38 | Gain |
|
| esv25440 | Chr.11:18714197 - 18716912 on Build GRCh38 | Loss |
|

Genomic Map