Assay Details
Target Gene Details
Entrez Gene ID: | 10380 |
Gene Name: | 3'(2'), 5'-bisphosphate nucleotidase 1 |
Gene Aliases: |
HEL20, PIP |
Location: |
Chr.1:220057482-220089916 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BPNT1 | NM_001286149.1 | 6 | 600 | NP_001273078.1 |
| NM_001286150.1 | 6 | 620 | NP_001273079.1 | |
| NM_001286151.1 | 6 | 642 | NP_001273080.1 | |
| NM_006085.5 | 7 | 728 | NP_006076.4 | |
| XM_005272998.2 | 8 | 744 | XP_005273055.1 | |
| XM_005272999.4 | 8 | 853 | XP_005273056.1 | |
| XM_005273000.4 | 7 | 777 | XP_005273057.1 | |
| XM_005273004.2 | XP_005273061.1 | |||
| XM_005273005.4 | XP_005273062.1 | |||
| XM_006711114.3 | 7 | 717 | XP_006711177.1 | |
| XM_011509063.2 | 7 | 691 | XP_011507365.1 | |
| XM_011509064.2 | 6 | 600 | XP_011507366.1 | |
| XM_017000043.1 | 6 | 692 | XP_016855532.1 | |
| XM_017000044.1 | 5 | 527 | XP_016855533.1 | |
| XM_017000046.1 | XP_016855535.1 | |||
| AF125042.1 | 7 | 665 | AAD17329.1 | |
| AJ249339.1 | 7 | 693 | CAB65115.1 | |
| AK021941.1 | 2 | 126 | ||
| AK291943.1 | 7 | 663 | ||
| AK298476.1 | 6 | 545 | ||
| AK300777.1 | 6 | 565 | ||
| AK315962.1 | 6 | 622 | ||
| BC017801.1 | 7 | 667 | AAH17801.1 | |
| EU794594.1 | 7 | 665 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832636 | Chr.1:219966355 - 220121010 on Build GRCh38 | Loss |
|
| nsv4632 | Chr.1:220041527 - 220076852 on Build GRCh38 | Insertion |
|
| nsv549222 | Chr.1:220053390 - 220073050 on Build GRCh38 | Loss |
|
| esv3588892 | Chr.1:220059332 - 220071714 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map