Assay Details
Target Gene Details
Entrez Gene ID: | 115701 |
Gene Name: | alpha kinase 2 |
Gene Aliases: |
HAK |
Location: |
Chr.18:58481247-58629192 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALPK2 | NM_052947.3 | 13 | 7209 | NP_443179.3 |
| XM_011525802.2 | 11 | 7829 | XP_011524104.1 | |
| XM_017025538.1 | 10 | 6327 | XP_016881027.1 | |
| XM_017025539.1 | 10 | 5075 | XP_016881028.1 | |
| AL832018.2 | 10 | 5541 | CAD89922.1 | |
| AY044450.1 | 10 | 5276 | AAK95952.1 | |
| BX647446.1 | 9 | 4687 | ||
| BX647639.1 | 10 | 6075 | ||
| BX647796.1 | 13 | 7196 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv577049 | Chr.18:58470593 - 58494881 on Build GRCh38 | Loss |
|
| dgv992e212 | Chr.18:58480806 - 58489401 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map