Assay Details
Target Gene Details
Entrez Gene ID: | 136 |
Gene Name: | adenosine A2b receptor |
Gene Aliases: |
ADORA2 |
Location: |
Chr.17:15927781-15975896 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADORA2B | NM_000676.2 | 1 | 371 | NP_000667.1 |
| XM_011523659.2 | XP_011521961.1 | |||
| XM_011523661.2 | 1 | 493 | XP_011521963.1 | |
| AY136748.1 | AAN01274.1 | |||
| BC025722.1 | 1 | 126 | AAH25722.1 | |
| M97759.1 | 1 | 174 | AAA51598.1 | |
| X68487.1 | 1 | 371 | CAA48505.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1061987 | Chr.17:15906703 - 15951821 on Build GRCh38 | Loss |
|
| esv2715702 | Chr.17:15750514 - 16668268 on Build GRCh38 | Deletion |
|
| dgv3107n100 | Chr.17:15703953 - 16150198 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73274391] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map