Assay Details
Target Gene Details
Entrez Gene ID: | 55190 |
Gene Name: | nudix hydrolase 11 |
Gene Aliases: |
APS1, ASP1, DIPP3b, DIPP3beta, hDIPP3beta |
Location: |
Chr.X:51490011-51496607 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDT11 | NM_018159.3 | 2 | 1219 | NP_060629.2 |
| AK001490.1 | 2 | 1106 | BAA91720.1 | |
| BC009942.2 | 2 | 1173 | AAH09942.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524351 | Chr.X:51454859 - 51498820 on Build GRCh38 | Gain |
|
| nsv471604 | Chr.X:51416463 - 51575197 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map