Assay Details
Target Gene Details
Entrez Gene ID: | 55002 |
Gene Name: | transmembrane and coiled-coil domains 3 |
Gene Aliases: |
C13orf11 |
Location: |
Chr.13:113490993-113550229 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1048814 | Chr.13:113081728 - 113611496 on Build GRCh38 | Gain |
|
| dgv586e212 | Chr.13:113497952 - 113505449 on Build GRCh38 | Loss |
|
| nsv1035248 | Chr.13:113454525 - 113505449 on Build GRCh38 | Loss |
|
| dgv585e212 | Chr.13:113492505 - 113504635 on Build GRCh38 | Loss |
|
| dgv376e214 | Chr.13:113502377 - 113522478 on Build GRCh38 | Loss |
|
| esv2759970 | Chr.13:113384114 - 113556749 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map