Assay Details
Target Gene Details
Entrez Gene ID: | 57795 |
Gene Name: | BMP/retinoic acid inducible neural specific 2 |
Gene Aliases: |
DBCCR1L2, FAM5B |
Location: |
Chr.1:177170922-177282422 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRINP2 | NM_021165.3 | 1 | 236 | NP_066988.1 |
| XM_005245379.1 | 1 | 702 | XP_005245436.1 | |
| AB051534.1 | 1 | 229 | ||
| AB161694.1 | 1 | 229 | BAD34946.1 | |
| AK296524.1 | ||||
| BC028036.1 | 1 | 128 | AAH28036.1 | |
| DA118275.1 | 1 | 236 |
More Information
Set Membership: |
|

Genomic Map