Assay Details
Target Gene Details
Entrez Gene ID: | 377 |
Gene Name: | ADP ribosylation factor 3 |
Gene Aliases: |
- |
Location: |
Chr.12:48928836-48957526 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARF3 | NM_001659.2 | 5 | 1468 | NP_001650.1 |
| XM_005268856.1 | 6 | 1782 | XP_005268913.1 | |
| XM_006719391.3 | XP_006719454.1 | |||
| AK291633.1 | ||||
| AK300080.1 | 2 | 665 | ||
| AK315970.1 | 2 | 598 | ||
| BC007647.1 | 5 | 1423 | AAH07647.1 | |
| BC017565.2 | 5 | 1346 | AAH17565.1 | |
| BC028402.2 | 5 | 1464 | AAH28402.1 | |
| M74491.1 | 5 | 1526 | AAB59425.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832404 | Chr.12:48864777 - 49064772 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map