Assay Details
Target Gene Details
Entrez Gene ID: | 5083 |
Gene Name: | paired box 9 |
Gene Aliases: |
STHAG3 |
Location: |
Chr.14:36657568-36677807 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAX9 | NM_006194.3 | 5 | 1869 | NP_006185.1 |
| AB248958.1 | 4 | 1312 | BAE79273.1 | |
| AK310339.1 | 5 | 754 | ||
| BC001159.1 | 4 | 1332 | AAH01159.1 | |
| U59628.1 | 3 | 1003 | AAD09487.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819749 | Chr.14:36676619 - 36676792 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map