Assay Details
Target Gene Details
Entrez Gene ID: | 3614 |
Gene Name: | inosine monophosphate dehydrogenase 1 |
Gene Aliases: |
IMPD, IMPD1, IMPDH-I, LCA11, RP10, sWSS2608 |
Location: |
Chr.7:128392277-128409988 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IMPDH1 | NM_000883.3 | NP_000874.2 | ||
| NM_001102605.1 | NP_001096075.1 | |||
| NM_001142576.1 | NP_001136048.1 | |||
| NM_001304521.1 | NP_001291450.1 | |||
| NM_183243.2 | NP_899066.1 | |||
| XM_017012168.1 | XP_016867657.1 | |||
| XM_017012169.1 | XP_016867658.1 | |||
| XM_017012170.1 | XP_016867659.1 | |||
| XM_017012171.1 | XP_016867660.1 | |||
| XM_017012172.1 | XP_016867661.1 | |||
| XM_017012173.1 | XP_016867662.1 | |||
| AK054667.1 | ||||
| AK092452.1 | ||||
| AK122994.1 | ||||
| AK131294.1 | BAD18464.1 | |||
| BC033622.1 | AAH33622.2 | |||
| BX406207.2 | ||||
| CD014008.1 | ||||
| CD014010.1 | ||||
| DA566139.1 | ||||
| DA980797.1 | ||||
| DC326380.1 | ||||
| J05272.1 | AAA36114.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv515737 | Chr.7:128403981 - 128410363 on Build GRCh38 | Loss |
|
| nsv831124 | Chr.7:128330292 - 128507856 on Build GRCh38 | Gain+Loss |
|
| esv2493565 | Chr.7:128345641 - 128413323 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map