Assay Details
Target Gene Details
Entrez Gene ID: | 5861 |
Gene Name: | RAB1A, member RAS oncogene family |
Gene Aliases: |
RAB1, YPT1 |
Location: |
Chr.2:65086854-65130301 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB1A | NM_004161.4 | 6 | 1698 | NP_004152.1 |
| NM_015543.1 | 4 | 1470 | NP_056358.1 | |
| XM_005264468.1 | 5 | 1407 | XP_005264525.1 | |
| AK055927.1 | 4 | 1313 | BAB71048.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819395 | Chr.2:65087568 - 65091059 on Build GRCh38 | Loss |
|
| nsv834246 | Chr.2:64981876 - 65176937 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map